Learn More
Cole-Parmer Cole-Parmer 6-watt UV Lamp with 365nm and 254nm Wavelength Light Tubes, 115VAC
Catalog No. 50200191 Shop All Cole-Parmer Products
Encompass_Preferred
Lightweight lamp lets you easily switch from one UV light source to another. A built-in reflector enhances UV illumination, while the durable, extruded metal housing features scratch-resistant powder coating.
Catalog No. 50-200-191 Supplier Cole-Parmer Supplier No. 9762024
Lightweight lamp lets you easily switch from one UV light source to another. A built-in reflector enhances UV illumination, while the durable, extruded metal housing features scratch-resistant powder coating.