Learn More
Perkin Elmer US LLC Tungsten Diode Array Detector Lamp for LC Series 200
Catalog No. 509044158
Encompass_Preferred
The Series 200 Photo Diode Array Detector provides true UV/VISdetection andhigh resolution spectral data. The excellentsignal-to-noise characteristics make itideally suited forlow-volume or low concentration samples.
Catalog No. 50-904-4158 Supplier Perkin Elmer US LLC Supplier No. N2922011
The Series 200 Photo Diode Array Detector provides true UV/VISdetection andhigh resolution spectral data. The excellentsignal-to-noise characteristics make itideally suited forlow-volume or low concentration samples.