Learn More
Perkin Elmer US LLC Deuterium Lamp Phase 2 for LC Series 200 Diode Array Detector
Catalog No. 509050160
Encompass_Preferred
Our extensive quality control and inspection process demands the very best quality sources. Choosing a PerkinElmer deuterium, tungsten or xenon source provides outstanding ultraviolet and true visible performance. Exceptional performance anywhere in the detector’s 190 – 700 nm wavelength range. Lamp changes are quick and easy due to a unique self-aligning lamp mount.
Catalog No. 50-905-0160 Supplier Perkin Elmer US LLC Supplier No. N2922046
Our extensive quality control and inspection process demands the very best quality sources. Choosing a PerkinElmer deuterium, tungsten or xenon source provides outstanding ultraviolet and true visible performance. Exceptional performance anywhere in the detector’s 190 – 700 nm wavelength range. Lamp changes are quick and easy due to a unique self-aligning lamp mount.