Learn More
Bulbtronics Inc TUV 30W 36" 30W UVC Ultraviolet Germicidal Lamp
SDP
Catalog No. NC1618098 Shop All Bulbtronics Inc. Products
Encompass_Preferred
Lamp, Germicidal; Bulbtronics; Philips; T8 shape; Linear double ended; Base: G13/medium BI pin; Length: 36 in. (91.4cm); Diameter: 1 in. (2.5cm); 30w; Spectral output: 254nm; UV output: 11.2w; Life: 8000hr.
Catalog No. NC1618098 Supplier Bulbtronics Inc Supplier No. 0000233
Lamp, Germicidal; Bulbtronics; Philips; T8 shape; Linear double ended; Base: G13/medium BI pin; Length: 36 in. (91.4cm); Diameter: 1 in. (2.5cm); 30w; Spectral output: 254nm; UV output: 11.2w; Life: 8000hr.