Learn More
Optimize Technologies Deuterium Lamp, Waters 2487, 2488, Alliance
SDP
Catalog No. 502006699
Encompass
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is designed to work with Waters 2487, 2488, and Alliance pump models and is equivalent to OEM part number WAS081142.
Catalog No. 50-200-6699 Supplier Optimize Technologies Supplier No. 393202825
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is designed to work with Waters 2487, 2488, and Alliance pump models and is equivalent to OEM part number WAS081142.