Learn More
Optimize Technologies Xenon Lamp, Waters 470, 474
SDP
Catalog No. 502006700
Encompass
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is designed to work with Waters 470 and 474 pump models and is equivalent to OEM part numbers WAT047447 & WAT052665.
Catalog No. 50-200-6700 Supplier Optimize Technologies Supplier No. 393203312
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is designed to work with Waters 470 and 474 pump models and is equivalent to OEM part numbers WAT047447 & WAT052665.