Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Binder Inc LAMP WHITE BIOLUX 965 230V/18W
SDP

Catalog No. NC2235849 Shop All BINDER Inc. Products
Encompass_Preferred

lamp bright white Biolux 965 230V / 18W

Catalog No. NC2235849 Supplier Binder Inc Supplier No. 50080076
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

lamp bright white Biolux 965 230V / 18W

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.