Learn More
Med Vet International Lamp with Stand Assembly
Catalog No. 503046817
Encompass_Preferred
All in one set includes: 150mL glass alcohol lamp with sturdy wooden base to prevent being knocked over and wire gauze stand for supporting your specimen for heating. Alcohol not included. Individually boxed,
Catalog No. 50-304-6817 Supplier Med Vet International Supplier No. 975318
All in one set includes: 150mL glass alcohol lamp with sturdy wooden base to prevent being knocked over and wire gauze stand for supporting your specimen for heating. Alcohol not included. Individually boxed,