Learn More
Enzo Life Sciences LAMP 1 monoclonal antibody (Ly1C6) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMEN001D
Encompass_Preferred
Clone: LY1C6. Host: Mouse. Isotype: IgG1. Immunogen: Rat liver lysosomal membrane preparation. UniProt: P14562. Source: Purified from ascites. Species Reactivity: Human, Rat, Hamster. Applications: ICC, IF, IP, WB, Electron microscopy. Recommended dilutions: Immunocytochemistry (1:100). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-824