Learn More
Cayman Chemical LAMP1CD107a RabbIt MoncLN 50uL
SDP
Catalog No. 502917461
Encompass_Preferred
An inhibitor of quorum sensing inhibits activation of the quorum-sensing regulator LuxR in aBgalactosidase reporter assay at 25 UM
Catalog No. 50-291-7461 Supplier Cayman Chemical Supplier No. 3807450 UL
An inhibitor of quorum sensing inhibits activation of the quorum-sensing regulator LuxR in aBgalactosidase reporter assay at 25 UM