Learn More
Cayman Chemical LAMP1CD107a RabbIt MoncLN
SDP
Catalog No. 502916068
Encompass_Preferred
A PD-1/PD-L1 interaction inhibitor (IC50 9 nM in a TR-FRET assay)
Catalog No. 50-291-6068 Supplier Cayman Chemical Supplier No. 38074100 UL
A PD-1/PD-L1 interaction inhibitor (IC50 9 nM in a TR-FRET assay)