Learn More
ACROBiosystems Recombinant Protein;Human LAMP1 / CD107a Protein;HEK293;100UG;LA1,LAMP1,CD107a,LAMP-1
Catalog No. 501658286
Encompass_Preferred
Recombinant Protein;Human LAMP1 / CD107a Protein;100UG;Human LAMP1, His Tag (LA1-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Met 382 (Accession # NP_005552).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.2 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-165-8286 Supplier ACROBiosystems Supplier No. LA1H5221100UG
Recombinant Protein;Human LAMP1 / CD107a Protein;100UG;Human LAMP1, His Tag (LA1-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Met 382 (Accession # NP_005552).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.2 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.