Learn More
ACROBiosystems Recombinant Protein;Human LAMP1 / CD107a Protein;HEK293;1MG;LA1,LAMP1,CD107a,LAMP-1
Supplier:  ACROBiosystems LA1H52211MG
Encompass_Preferred
Recombinant Protein;Human LAMP1 / CD107a Protein;1MG;Human LAMP1, His Tag (LA1-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Met 382 (Accession # NP_005552).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.2 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-165-8287