Learn More
Enzo Life Sciences LAMP2 monoclonal antibody (GL2A7) (200µl)
SDP
Supplier:  Enzo Life Sciences ADIVAMEN003F
Encompass_Preferred
Clone: GL2A7. Host: Rat. Isotype: IgG. Immunogen: Mouse liver lysosomal membrane preparation. UniProt ID: P17047. Source: Purified from ascites. Species reactivity: Mouse Rabbit. Applications: ICC, IP, WB. Recommended Dilutions: Immunocytochemistry (1:100) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. Rat Ascites. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7684