Learn More
Enzo Life Sciences LAMP2 monoclonal antibody (GL2A7) (50µl)
SDP
Supplier:  Enzo Life Sciences ADIVAMEN003D
Encompass_Preferred
Clone: GL2A7. Host: Rat. Isotype: IgG. Immunogen: Mouse liver lysosomal membrane preparation. Source: Purified from ascites. Species reactivity: Mouse Rabbit. Applications: ICC, IP, WB. Formulation: Liquid. Rat Ascites.
Catalog No. 50-200-7683