Learn More
Enzo Life Sciences LAMP3 (human) monoclonal antibody (104G4) (Alexa Fluor 546) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS1850100
Encompass_Preferred
Clone: 104G4. Isotype: IgG1. Immunogen: Human DC-LAMP alkaline phosphatase fusion protein. Species reactivity: Human Sheep. Specificity: Recognizes an epitope in the intracytoplasmic domain of DC-LAMP. Applications: IF, IHC (FS). Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-201-2737