Learn More
Grainger Lampholder INC Surface Mnt Black 1 Head
Catalog No. 502749337
Encompass_Preferred
The W.F. Harris Lighting 14G273 is an outdoor lampholder designed for surface mounting applications. It features a cast aluminum construction with a gray powder coat finish and stainless steel hardware. The lampholder has a medium base socket that accepts up to a 100W incandescent bulb. It is UL and CUL listed for wet locations. The dimensions are 5-1/2" high x 5" wide x 4-3/4" deep. This outdoor lampholder provides a durable and weather-resistant lighting fixture solution for a variety of exterior lighting needs.
Catalog No. 50-274-9337 Supplier Grainger Supplier No. 14G273
The W.F. Harris Lighting 14G273 is an outdoor lampholder designed for surface mounting applications. It features a cast aluminum construction with a gray powder coat finish and stainless steel hardware. The lampholder has a medium base socket that accepts up to a 100W incandescent bulb. It is UL and CUL listed for wet locations. The dimensions are 5-1/2" high x 5" wide x 4-3/4" deep. This outdoor lampholder provides a durable and weather-resistant lighting fixture solution for a variety of exterior lighting needs.