Learn More
Grainger Lampholder INC Surface Mount Bronze
Catalog No. 502750342
Encompass_Preferred
The W.F. Harris Lighting 14G262 is an outdoor lampholder designed for surface mounting applications. It features a porcelain socket with nickel plated brass shell and backplate. The lampholder accepts medium base bulbs up to 300W. It has a pull chain socket with 8" lead wires and is UL and CUL listed for wet locations. The dimensions are 4-1/2" diameter x 2-3/4" projection. This surface mount lampholder provides a durable and weatherproof socket for outdoor lighting fixtures.
Catalog No. 50-275-0342 Supplier Grainger Supplier No. 14G262
The W.F. Harris Lighting 14G262 is an outdoor lampholder designed for surface mounting applications. It features a porcelain socket with nickel plated brass shell and backplate. The lampholder accepts medium base bulbs up to 300W. It has a pull chain socket with 8" lead wires and is UL and CUL listed for wet locations. The dimensions are 4-1/2" diameter x 2-3/4" projection. This surface mount lampholder provides a durable and weatherproof socket for outdoor lighting fixtures.