Learn More
Analytical Sales & Services Lancer C18 Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID
Catalog No. 5000805683
Encompass
Lancer C18 Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID
Catalog No. 50-008-05683 Supplier Analytical Sales & Services Supplier No. ADV5077
Lancer C18 Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID