Learn More
Analytical Sales & Services Lancer C18 Analytical Column. 120Â, 5µm, 10cm x 3.0mm ID
Catalog No. 5000805759
Encompass
Lancer C18 Analytical Column. 120Â, 5µm, 10cm x 3.0mm ID
Catalog No. 50-008-05759 Supplier Analytical Sales & Services Supplier No. ADV5057
Lancer C18 Analytical Column. 120Â, 5µm, 10cm x 3.0mm ID