Learn More
Analytical Sales & Services Lancer C18 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID
Catalog No. 5000806709
Encompass
Lancer C18 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID
Catalog No. 50-008-06709 Supplier Analytical Sales & Services Supplier No. ADV5011
Lancer C18 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID