Learn More
Analytical Sales & Services Lancer C18 Analytical Column. 120Â, 5µm, 15cm x 4.6mm ID
Catalog No. 5000805760
Encompass
Lancer C18 Analytical Column. 120Â, 5µm, 15cm x 4.6mm ID
Catalog No. 50-008-05760 Supplier Analytical Sales & Services Supplier No. ADV5012
Add to Cart
Add to Cart
Lancer C18 Analytical Column. 120Â, 5µm, 15cm x 4.6mm ID