Learn More
Analytical Sales & Services Lancer C18 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID
Catalog No. 5000806710
Encompass
Lancer C18 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID
Catalog No. 50-008-06710 Supplier Analytical Sales & Services Supplier No. ADV5059
Add to Cart
Add to Cart
Lancer C18 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID