Learn More
Analytical Sales & Services Lancer C18 Analytical Column. 120Â, 5µm, 25cm x 4.6mm ID
Catalog No. 5000805761
Encompass
Lancer C18 Analytical Column. 120Â, 5µm, 25cm x 4.6mm ID
Catalog No. 50-008-05761 Supplier Analytical Sales & Services Supplier No. ADV5013
Lancer C18 Analytical Column. 120Â, 5µm, 25cm x 4.6mm ID