Learn More
Analytical Sales & Services Lancer C18 Analytical Column. 120Â, 5µm, 3cm x 4.6mm ID
Catalog No. 5000806708
Encompass
Lancer C18 Analytical Column. 120Â, 5µm, 3cm x 4.6mm ID
Catalog No. 50-008-06708 Supplier Analytical Sales & Services Supplier No. ADV5009
Lancer C18 Analytical Column. 120Â, 5µm, 3cm x 4.6mm ID