Learn More
Analytical Sales & Services Lancer C18 Microbore Column. 120Â, 5µm, 10cm x 0.3mm ID
Catalog No. 5000804577
Encompass
Lancer C18 Microbore Column. 120Â, 5µm, 10cm x 0.3mm ID
Catalog No. 50-008-04577 Supplier Analytical Sales & Services Supplier No. ADV10L3L205
Lancer C18 Microbore Column. 120Â, 5µm, 10cm x 0.3mm ID