Learn More
Analytical Sales & Services Lancer C18 Microbore Column. 120Â, 5µm, 10cm x 1.0mm ID
Catalog No. 5000805952
Encompass
Lancer C18 Microbore Column. 120Â, 5µm, 10cm x 1.0mm ID
Catalog No. 50-008-05952 Supplier Analytical Sales & Services Supplier No. ADV10L1L205
Lancer C18 Microbore Column. 120Â, 5µm, 10cm x 1.0mm ID