Learn More
Analytical Sales & Services Lancer C18 Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID
Catalog No. 5000805900
Encompass
Lancer C18 Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID
Catalog No. 50-008-05900 Supplier Analytical Sales & Services Supplier No. ADV05L5L205
Add to Cart
Add to Cart
Lancer C18 Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID