Learn More
Analytical Sales & Services Lancer C18 PEEK Guard Cartridge. 5µm, 20mm x 2.1mm ID
Catalog No. 5000805628
Encompass
Lancer C18 PEEK Guard Cartridge. 5µm, 20mm x 2.1mm ID
Catalog No. 50-008-05628 Supplier Analytical Sales & Services Supplier No. ADVH185
Lancer C18 PEEK Guard Cartridge. 5µm, 20mm x 2.1mm ID