Learn More
Analytical Sales & Services Lancer C18 Prep Column. 120Â, 10µm, 10cm x 20mm ID
Catalog No. 5000805622
Encompass
Lancer C18 Prep Column. 120Â, 10µm, 10cm x 20mm ID
Catalog No. 50-008-05622 Supplier Analytical Sales & Services Supplier No. ADV601020
Lancer C18 Prep Column. 120Â, 10µm, 10cm x 20mm ID