Learn More
Analytical Sales & Services Lancer C18 Prep Column. 120Â, 10µm, 15cm x 20mm ID
Catalog No. 5000806087
Encompass
Lancer C18 Prep Column. 120Â, 10µm, 15cm x 20mm ID
Catalog No. 50-008-06087 Supplier Analytical Sales & Services Supplier No. ADV601520
Lancer C18 Prep Column. 120Â, 10µm, 15cm x 20mm ID