Learn More
Analytical Sales & Services Lancer C18 Prep Column. 120Â, 10um, 25cm x 20mm ID
Catalog No. 5000805623
Encompass
Lancer C18 Prep Column. 120Â, 10um, 25cm x 20mm ID
Catalog No. 50-008-05623 Supplier Analytical Sales & Services Supplier No. ADV602520
Lancer C18 Prep Column. 120Â, 10um, 25cm x 20mm ID