Learn More
Analytical Sales & Services Lancer C18 Prep Column. 120Â, 5µm, 25cm x 20mm ID
Catalog No. 5000805998
Encompass
Lancer C18 Prep Column. 120Â, 5µm, 25cm x 20mm ID
Catalog No. 50-008-05998 Supplier Analytical Sales & Services Supplier No. ADV532520
Lancer C18 Prep Column. 120Â, 5µm, 25cm x 20mm ID