Learn More
Analytical Sales & Services Lancer C18 Prep Guard Cartridge. 120Â, 10µm, 25mm x 20mm ID
Catalog No. 5000806134
Encompass
Lancer C18 Prep Guard Cartridge. 120Â, 10µm, 25mm x 20mm ID
Catalog No. 50-008-06134 Supplier Analytical Sales & Services Supplier No. ADV0220181
Lancer C18 Prep Guard Cartridge. 120Â, 10µm, 25mm x 20mm ID