Learn More
Analytical Sales & Services Lancer C18 Semi-Prep Column. 120Â, 10µm, 15cm x 10mm ID
Catalog No. 5000805624
Encompass
Lancer C18 Semi-Prep Column. 120Â, 10µm, 15cm x 10mm ID
Catalog No. 50-008-05624 Supplier Analytical Sales & Services Supplier No. ADV601510
Add to Cart
Add to Cart
Lancer C18 Semi-Prep Column. 120Â, 10µm, 15cm x 10mm ID