Learn More
Analytical Sales & Services Lancer C18 Semi-Prep Column. 120Â, 5µm, 10cm x 10mm ID
Catalog No. 5000806715
Encompass
Lancer C18 Semi-Prep Column. 120Â, 5µm, 10cm x 10mm IDQty g:
Catalog No. 50-008-06715 Supplier Analytical Sales & Services Supplier No. ADV531010
Lancer C18 Semi-Prep Column. 120Â, 5µm, 10cm x 10mm IDQty g: