Learn More
Analytical Sales & Services Lancer C18 Semi-Prep Column. 120Â, 5µm, 25cm x 10mm ID
Catalog No. 5000806152
Encompass
Lancer C18 Semi-Prep Column. 120Â, 5µm, 25cm x 10mm ID
Catalog No. 50-008-06152 Supplier Analytical Sales & Services Supplier No. ADV532510
Lancer C18 Semi-Prep Column. 120Â, 5µm, 25cm x 10mm ID