Learn More
Analytical Sales & Services Lancer C18 Semi-Prep SS Guard Cartridge. 10µm, 3cm x 10mm ID
Catalog No. 5000804986
Encompass
Lancer C18 Semi-Prep Stainless Steel Guard Cartridge. 10µm, 30mm x 10mm ID
Catalog No. 50-008-04986 Supplier Analytical Sales & Services Supplier No. ADV0310181
Lancer C18 Semi-Prep Stainless Steel Guard Cartridge. 10µm, 30mm x 10mm ID