Learn More
Analytical Sales & Services Lancer C8 Analytical Column. 120Â, 4.5µm, 10cm x 2.1mm ID
Catalog No. 5000805606
Encompass
Lancer C8 Analytical Column. 120Â, 4.5µm, 10cm x 2.1mm ID
Catalog No. 50-008-05606 Supplier Analytical Sales & Services Supplier No. ADV5277
Lancer C8 Analytical Column. 120Â, 4.5µm, 10cm x 2.1mm ID