Learn More
Analytical Sales & Services Lancer C8 Analytical Column. 120Â, 4.5µm, 25cm x 4.6mm ID
Catalog No. 5000805602
Encompass
Lancer C8 Analytical Column. 120Â, 4.5µm, 25cm x 4.6mm ID
Catalog No. 50-008-05602 Supplier Analytical Sales & Services Supplier No. ADV5213
Lancer C8 Analytical Column. 120Â, 4.5µm, 25cm x 4.6mm ID