Learn More
Analytical Sales & Services Lancer C8 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID
Catalog No. 5000805966
Encompass
Lancer C8 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID
Catalog No. 50-008-05966 Supplier Analytical Sales & Services Supplier No. ADV5211
Lancer C8 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID