Learn More
Analytical Sales & Services Lancer C8 Analytical Column. 120Â, 5µm, 15cm x 2.1mm ID
Catalog No. 5000806660
Encompass
Lancer C8 Analytical Column. 120Â, 5µm, 15cm x 2.1mm ID
Catalog No. 50-008-06660 Supplier Analytical Sales & Services Supplier No. ADV5278
Lancer C8 Analytical Column. 120Â, 5µm, 15cm x 2.1mm ID