Learn More
Analytical Sales & Services Lancer C8 Analytical Column. 120Â, 5µm, 25cm x 2.1mm ID
Catalog No. 5000806082
Encompass
Lancer C8 Analytical Column. 120Â, 5µm, 25cm x 2.1mm ID
Catalog No. 50-008-06082 Supplier Analytical Sales & Services Supplier No. ADV5279
Lancer C8 Analytical Column. 120Â, 5µm, 25cm x 2.1mm ID