Learn More
Analytical Sales & Services Lancer C8 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID
Supplier:  Analytical Sales & Services ADV5259
Encompass
Lancer C8 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID
Catalog No. 50-008-06712