Learn More
Analytical Sales & Services Lancer C8 Analytical Column. 120Â, 5µm, 5cm x 2.1mm ID
Catalog No. 5000805477
Encompass
Lancer C8 Analytical Column. 120Â, 5µm, 5cm x 2.1mm ID
Catalog No. 50-008-05477 Supplier Analytical Sales & Services Supplier No. ADV5276
Lancer C8 Analytical Column. 120Â, 5µm, 5cm x 2.1mm ID