Learn More
Analytical Sales & Services Lancer C8 Analytical Column. 120Â, 5µm, 5cm x 3.0mm ID
Catalog No. 5000805446
Encompass
Lancer C8 Analytical Column. 120Â, 5µm, 5cm x 3.0mm ID
Catalog No. 50-008-05446 Supplier Analytical Sales & Services Supplier No. ADV5256
Lancer C8 Analytical Column. 120Â, 5µm, 5cm x 3.0mm ID