Learn More
Analytical Sales & Services Lancer C8 Microbore Column. 120Â, 5µm, 15cm x 0.5mm ID
Catalog No. 5000806645
Encompass
Lancer C8 Microbore Column. 120Â, 5µm, 15cm x 0.5mm ID
Catalog No. 50-008-06645 Supplier Analytical Sales & Services Supplier No. ADV15L5L000
Lancer C8 Microbore Column. 120Â, 5µm, 15cm x 0.5mm ID