Learn More
Analytical Sales & Services Lancer C8 Microbore Column. 120Â, 5µm, 25cm x 1.0mm ID
Catalog No. 5000806646
Encompass
Lancer C8 Microbore Column. 120Â, 5µm, 25cm x 1.0mm ID
Catalog No. 50-008-06646 Supplier Analytical Sales & Services Supplier No. ADV25L1L000
Lancer C8 Microbore Column. 120Â, 5µm, 25cm x 1.0mm ID