Learn More
Analytical Sales & Services Lancer C8 Microbore Column. 120Â, 5µm, 3cm x 0.3mm ID
Catalog No. 5000805883
Encompass
Lancer C8 Microbore Column. 120Â, 5µm, 3cm x 0.3mm ID
Catalog No. 50-008-05883 Supplier Analytical Sales & Services Supplier No. ADV03L3L000
Lancer C8 Microbore Column. 120Â, 5µm, 3cm x 0.3mm ID