Learn More
Analytical Sales & Services Lancer C8 Microbore Column. 120Â, 5µm, 3cm x 1.0mm ID
Catalog No. 5000805880
Encompass
Lancer C8 Microbore Column. 120Â, 5µm, 3cm x 1.0mm ID
Catalog No. 50-008-05880 Supplier Analytical Sales & Services Supplier No. ADV03L1L000
Add to Cart
Add to Cart
Lancer C8 Microbore Column. 120Â, 5µm, 3cm x 1.0mm ID